Learn More
Antimony potassium tartrate trihydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04061922
Description
Antimony potassium tartrate trihydrate is used as a antiparasitic agent, which used in the treatment of schistosomiasis and leishmaniasis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Antimony potassium tartrate trihydrate | |
Powder | |
99.0-103.0% | |
MFCD00148863 | |
Soluble in water and glycerine. Insoluble in ethanol. | |
ACS |
28300-74-5 | |
100 g | |
C8H4K2O12Sb2·3H2O | |
Potassium antimonyl tartrate | |
667.87 (613.82 Anhydrous) | |
2.6 g/cm3 |
Chemical Identifiers
28300-74-5 | |
C8H4K2O12Sb2·3H2O | |
MFCD00148863 | |
Potassium antimonyl tartrate |
Safety and Handling
Avoid release to the environment. Refer to special instructions/Safety data sheets
P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c
H302+H332-H335-H373
DOTInformation : DOT Class: 6.1, Packaging Group: III
EINECSNumber : 229-436-1
RTECSNumber : CC6825000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only