missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony potassium tartrate trihydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

$57.41 - $604.34

Chemical Identifiers

CAS 28300-74-5
Molecular Formula C8H4K2O12Sb2·3H2O
MDL Number MFCD00148863
Synonym Potassium antimonyl tartrate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4061914
View Documents
Thermo Scientific Chemicals
04061914
25 g
Each for $57.41
 
AA4061922
View Documents
Thermo Scientific Chemicals
04061922
100 g
Each for $139.08
 
AA4061936
View Documents
Thermo Scientific Chemicals
04061936
500 g
Each for $604.34
 
Description

Description

Antimony potassium tartrate trihydrate is used as a antiparasitic agent, which used in the treatment of schistosomiasis and leishmaniasis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

28300-74-5
C8H4K2O12Sb2·3H2O
MFCD00148863
Potassium antimonyl tartrate

Specifications

28300-74-5
25 g
C8H4K2O12Sb2·3H2O
Potassium antimonyl tartrate
667.87 (613.82 Anhydrous)
2.6
Powder
99.0-103.0%
MFCD00148863
Soluble in water and glycerine. Insoluble in ethanol.
ACS
Antimony potassium tartrate trihydrate
Videos
Safety and Handling

Safety and Handling

Avoid release to the environment. Refer to special instructions/Safety data sheets

P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c

H302+H332-H335-H373

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 229-436-1

RTECSNumber : CC6825000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only