Promega ApoTox-Glo™ Triplex Assay System DFS Item
The ApoTox-Glo™ Triplex Assay combines three assay chemistries to easily assess viability, cytotoxicity and apoptosis events in the same cell-based assay well.
$1152.91 - $4074.01
Specifications
Content And Storage -30°C to -10°C
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRG6320 DFS Item
View Documents
Promega
G6320
10 mL
Each for $1,152.91
PRG6321 DFS Item
View Documents
Promega
G6321
5 x 10 mL
Each for $4,074.01
Description
Description
  • Assess Viability, Cytotoxicity and Apoptosis in One Assay
  • Easy to implement with an add-mix-measure protocol
  • Normalize data with a built-in control
  • Flexible and easily automated
Specifications
Specifications
-30°C to -10°C
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
  • (a)U.S. Pat. Nos. 7,416,854, 7,553,632 and other patents pending.
  • (b)U.S. Pat. Nos. 6,602,677 and 7,241,584, Australian Pat. Nos. 754312 and 785294 and other patents and patents pending.
  • (c)The method of recombinant expression of Coleoptera luciferase is covered by U.S. Pat. Nos. 5,583,024, 5,674,713 and 5,700,673.