Learn More
Promega ApoTox-Glo™ Triplex Assay System
The ApoTox-Glo™ Triplex Assay combines three assay chemistries to easily assess viability, cytotoxicity and apoptosis events in the same cell-based assay well.
Supplier: Promega G6320
Description
- Assess Viability, Cytotoxicity and Apoptosis in One Assay
- Easy to implement with an add-mix-measure protocol
- Normalize data with a built-in control
- Flexible and easily automated
Specifications
| 10 mL | |
| 10 mL Assay Buffer, 10 μL GF-AFC substrate, 10 μL bis-AAF-R110 substrate, 10 mL Caspase-Glo™ 3/7 Buffer, 1 bottle Caspase-Glo 3/7 substrate | |
| -30°C to -10°C |
- (a)U.S. Pat. Nos. 7,416,854, 7,553,632 and other patents pending.
- (b)U.S. Pat. Nos. 6,602,677 and 7,241,584, Australian Pat. Nos. 754312 and 785294 and other patents and patents pending.
- (c)The method of recombinant expression of Coleoptera luciferase is covered by U.S. Pat. Nos. 5,583,024, 5,674,713 and 5,700,673.