missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Applied Biosystems™ MicroAmp™ Optical 96-Well Reaction Plate DFS Item

Designed to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification

$116.00 - $5210.00

Specifications

Barcode No Barcode
Certifications/Compliance Certified DNA/RNase Free
Product Line MicroAmp
Color Optical
Product Type Microplate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N8010560 DFS Item
View Documents Promotion Details
Applied Biosystems™
N8010560
10 Plates
Each for $116.00
 
4316813 DFS Item
View Documents
Applied Biosystems™
4316813
500 Plates
Each for $5,210.00
 
Description

Description

This plate, constructed from a single rigid piece of polypropylene in a 96-well format, is compatible with Applied Biosystems 96-well Real-Time PCR systems and thermal cyclers.

  • Frosted to minimize interfering fluorescence from wells of cycling block
  • Screened to eliminate auto-fluorescent plates
  • Unique design provides barrier to ambient air to help ensure well-to-well temperature uniformity
  • Capacity: 96 x 0.2mL tubes

  • Platform: 2720 Thermal Cycler, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 6100 Nucleic Acid PrepStation, 7000 System, 7300 System, 7500 System, 7900HT System, GeneAmp™ 9700, ProFlex™ PCR System, QuantStudio™, QuantStudio Dx, SimpliAmp™ Thermal Cycler, Veriti™ Dx Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA 7 System

Applications: PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

Specifications

Specifications

No Barcode
MicroAmp
Microplate
Semi
96
Certified DNA/RNase Free
Optical
Thermal Cycler, Genetic Analyzer, DNA Analyzer
Polypropylene
0.2 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'