Fisherbrand™ Trays and Dividers for Shell Vials
Accommodate narrow- or wide-diameter vials.
 View more versions of this product
Catalog No. AS261
Explore available promotions
Description
Description
  • Cardboard trays have cell dividers
  • 10 × 10 in. (25 x 25cm) row configuration
Specifications
Specifications
Dividers
Cardboard
Not Autoclavable
Tray for Wide Vials
For wide vials (28.5 x 95 mm)