missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Trays and Dividers for Shell Vials
Accommodate narrow- or wide-diameter vials.
$372.45 - $421.97
Catalog Number | Mfr. No. | Type | For Use With (Equipment) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | For Use With (Equipment) | Price | Quantity | ||||
AS261
|
N/A | Tray for Wide Vials | For wide vials (28.5 x 95 mm) |
Case of 50 Each for $421.97
|
|
||||
AS260
|
N/A | Tray for Narrow Vials | For narrow vials (25 x 95 mm) |
Case of 50 Each for $372.45
|
|
||||
Description
- Cardboard trays have cell dividers
- 10 x 10 in. (25 x 25cm) row configuration
Specifications
Not Autoclavable | |
Cardboard | |
Dividers |