National Safety Apparel™ ArcGuard™ 20 Cal Short Coat Kit (11)
Arc flash kit with Purview FR short coat and bib overall in Ultrasoft
Supplier:  National Safety Apparel™ KIT2SC205X11
 View more versions of this product
Catalog No. 170011332
Explore available promotions
Description
Description
  • Arc Rating: 20 cal/cm²
  • CAT 2
  • Fabric/Material: 13 oz. Westex™ UltraSoft™
  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • FR hook and loop front closure on short coat
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Hook and loop at side of bib for easy access
  • Expandable leg opening to fit over boots
  • Made in the USA
Specifications
Specifications
20 cal/cm²
CAT 2, NFPA 70 E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Westex™ UltraSoft™
FR Hook and Loop
Adjustable
Navy
Open
5X-Large
Stitched