Thermo Scientific™ ART™ Pipette Tip Empty Hinged Racks
Save valuable space in the lab with these stackable pipette tip hinged racks.
Supplier:  Thermo Scientific™ 2008ER
 View more versions of this product
Catalog No. 02-682-272
Explore available promotions
Description
Description
Save valuable space in the lab with the stackable Thermo Scientific™ ART™ Pipette Tip Empty Hinged Racks. ART barrier tips provide 100% security against aerosols and liquid contamination. Front latch closes and locks, ensuring safe and secure storage throughout.
Features:
  • ART barrier tips provide 100% security against aerosols and liquid contamination
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
Certifications:
  • Nonsterile products certified to be free of RNase, DNase, DNA, ATP, Endotoxin, Pyrogen and PCR Inhibitors
  • Sterile products certified to be free of RNase, DNase, DNA, ATP, Bioburden, Endotoxin, Pyrogen and PCR Inhibitors
Recommended for:
  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Specifications
Specifications
Empty Hinged Rack
Square
Non-sterile
Hinged Rack, 8 trays/pack, 5 packs/case
Large
Yes
Empty Hinged Racks
Hinged Rack, 8 trays/Pk., 5 Pks,/Cs.