missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Medicom™ AssureTouch™ Power™ Nitrile Medical Examination Gloves with Textured Fingertips
AssureTouch™ Power™ Nitrile Medical Examination Gloves with Textured Fingertips. Provides strong puncture and tear resistance without compromising tactility. Extended cuffs provide optimal coverage. Textured fingertips for a secure grip, even when wet. Chemo tested and fentanyl safe.
Supplier: Medicom™ 9999B
Description
- These gloves provide strong puncture, tear and chemical resistance without compromising dexterity or tactility.
- Extended cuffs provide optimal coverage.
- Textured fingertips ensure a secure grip, even when wet.
- Chemo tested and fentanyl safe.
Specifications
No | |
Healthcare, Industrial, Laboratory | |
Blue | |
Fingertip Textured | |
Powder Free | |
Non-sterile | |
Beaded |
No | |
1.5 | |
Single-use | |
Yes | |
Small | |
Yes |