Fisherbrand™ ASTM™ Hydrometers, Plain Form
API hydrometers are available in regular and ASTM™ recommended forms.
$49.69 - $790.48
Catalog Number | Mfr. No. | ASTM Number | Certifications/Compliance | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | ASTM Number | Certifications/Compliance | Price | Quantity | ||||
11-603EC
|
N/A | 7H | 3-point Certification, NIST, Cited in ASTM E100-14 Table 1, ASTM E126 |
Each for $735.52
|
|
||||
11-603F
|
N/A | 8H | NIST, Cited in ASTM E100-14 Table 1, ASTM E126 |
Case for $50.00
|
|
||||
11-603AC
|
N/A | 3H | 3-point Certification, NIST, Cited in ASTM E100-14 Table 1, ASTM E126 |
Each for $735.34
|
|
||||
11-603E
|
N/A | 7H | NIST, Cited in ASTM E100-14 Table 1, ASTM E126 |
Each for $50.00
|
|
||||
11-603D
|
N/A | 6H | NIST, Cited in ASTM E100-14 Table 1, ASTM E126 |
Each for $50.14
|
|
||||
11-603A
|
N/A | 3H | NIST, Cited in ASTM E100-14 Table 1, ASTM E126 |
Each for $53.14
|
|
||||
11-603CC
|
N/A | 5H | 3-point Certification, NIST, Cited in ASTM E100-14 Table 1, ASTM E126 |
Each for $790.48
|
|
||||
11-603C
|
N/A | 5H | NIST, Cited in ASTM E100-14 Table 1, ASTM E126 |
Each for $50.00
|
|
||||
11-603H
|
N/A | 10H | NIST, Cited in ASTM E100-14 Table 1, ASTM E126 |
Each for $51.62
|
|
||||
11-603BC
|
N/A | 4H | 3-point Certification, NIST, Cited in ASTM E100-14 Table 1, ASTM E126 |
Each for $735.34
|
|
||||
11-603K
|
N/A | 2H | NIST, Cited in ASTM E100-14 Table 1, ASTM E126 |
Each for $50.14
|
|
||||
11-603B
|
N/A | 4H | NIST, Cited in ASTM E100-14 Table 1, ASTM E126 |
Each for $50.38
|
|
||||
11-603G
|
N/A | 9H | NIST, Cited in ASTM E100-14 Table 1, ASTM E126 |
Each for $49.69
|
|
||||
11-603DC
|
N/A | 6H | 3-point Certification, NIST, Cited in ASTM E100-14 Table 1, ASTM E126 |
Each for $735.79
|
|
||||
11-603KC
|
N/A | 2H | 3-point Certification, NIST, Cited in ASTM E100-14 Table 1, ASTM E126 |
Each for $735.69
|
|
||||
Description
These hydrometers are manufactured to meet or exceed ASTM™ E 100 specifications which are accepted by NIST™ and U.S. Bureau of Mines.
- Permanently marked with individual serial numbers
- Without thermometer in body
- Approximate length: 330mm
- Temperature of standardization: 60°F
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Certifications
A certificate is provided to indicate instrument traceability to standards provided by NIST.
Compliance
Meet or exceed ASTM E 100 specifications which are accepted by NIST and U.S. Bureau of Mines, Calibrated according to ASTM E 126 specifications.
Specifications
0.1° | |
330 mm | |
60°F |