Eisco™ ASTM Low Form Glass Beaker
Made from high quality borosilicate 3.3 glass. Extremely durable, with a low temperature gradient, and superior chemical resistance. Features a spout for easy pouring and graduations as per ASTM-E960.
$5.79 - $182.88
Specifications
Color | Clear |
---|---|
Material | Borosilicate Glass |
Certifications/Compliance | ASTM |
Catalog Number | Mfr. No. | Capacity (Metric) | No. per Pack | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | No. per Pack | Price | Quantity | ||||
S40869
|
Eisco™
CH0124C |
100 mL | 1 |
Each for $6.94
|
|
||||
S27513
|
Eisco™
CH0124A10PK12 |
10 mL | 12 |
Pack of 12 Each for $49.32
|
|
||||
S40872
|
Eisco™
CH0124CPK12 |
100 mL | 12 |
Each for $70.56
|
|
||||
S27514
|
Eisco™
CH0124A20PK12 |
20 mL | 12 |
Pack of 12 Each for $56.35
|
|
||||
S40873
|
Eisco™
CH0124E |
250 mL | 1 |
Pack for $10.76
|
|
||||
S27515
|
Eisco™
CH0124A30PK12 |
30 mL | 12 |
Pack of 12 Each for $56.35
|
|
||||
S40874
|
Eisco™
CH0124EPK12 |
250 mL | 12 |
Pack for $109.15
|
|
||||
S27516
|
Eisco™
CH0124B |
50 mL | 1 |
Each for $5.79
|
|
||||
S40875
|
Eisco™
CH0124F |
400 mL | 1 |
Pack for $15.85
|
|
||||
S27517
|
Eisco™
CH0124D |
150 mL | 1 |
Each for $7.65
|
|
||||
S40880
|
Eisco™
CH0124G |
600 mL | 1 |
Each for $17.15
|
|
||||
S27518
|
Eisco™
CH0124FPK6 |
400 mL | 6 |
Pack of 6 Each for $87.15
|
|
||||
S40882
|
Eisco™
CH0124GPK6 |
600 mL | 6 |
Each for $102.74
|
|
||||
S27519
|
Eisco™
CH0124J |
2000 mL | 1 |
Each for $42.94
|
|
||||
S40884
|
Eisco™
CH0124I |
1000 mL | 1 |
Each for $30.53
|
|
||||
S40886
|
Eisco™
CH0124IPK6 |
1000 mL | 6 |
Each for $182.88
|
|
||||
Description
Warranty
1 year limited manufacturer warranty
Specifications
Clear | |
Borosilicate Glass | |
ASTM |