Eisco™ Augen Gneiss Specimen (Metamorphic Rock)
For use in learning about differential stresses and foliation in metamorphic rocks. EISCO Augen Gneiss Specimen (Metamorphic Rock) is high-grade metamorphic rock formed from other rocks undergoing alteration from intense heat and pressure.
Supplier:  Eisco™ ESNG0054
 View more versions of this product
Catalog No. S24778
Explore available promotions
Description
Description
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Relict xenocrysts in sample record direction of stress during alteration
  • This rock sample is great for demonstrating metamorphic properties and can be used in lessons about rock cycle
  • Comes in clear labeled and barcoded bag for storage
Specifications
Specifications
Clear labeled and barcoded bag
Learning about differential stresses and foliation in metamorphic rocks
Approx. 1 in.
Each
Augen Gneiss Study Specimen (Metamorphic Rock)
Approx. 3 cm