Aushon Chemiluminescent Human Chemokine Array Kits DFS Item
For simultaneous measurement of combinations of chemokines
Supplier:  Aushon Biosystems Inc 030381FA
 View more versions of this product
Catalog No. 030381FA
Explore available promotions
Description
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Specifications
Human
For simultaneous measurement of combinations of chemokines
Chemokine Array Kits
3 Plex