missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aushon Chemiluminescent Human Chemokine Array Kits
For simultaneous measurement of combinations of chemokines
$876.65 - $1850.71
Specifications
Reactivity | Human |
---|---|
For Use With (Application) | For simultaneous measurement of combinations of chemokines |
Product Type | Chemokine Array Kits |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
030041FA
|
Aushon Biosystems Inc
030041FA |
1 Plex |
Each for $876.65
|
|
|||||
030011FA
|
Aushon Biosystems Inc
030011FA |
1 Plex |
Each for $876.65
|
|
|||||
030021FA
|
Aushon Biosystems Inc
030021FA |
1 Plex |
Each for $876.65
|
|
|||||
030381FA
|
Aushon Biosystems Inc
030381FA |
3 Plex |
Each for $1,266.29
|
|
|||||
03F801FA
|
Aushon Biosystems Inc
03F801FA |
5 Plex |
Each for $1,655.90
|
|
|||||
03FC01FA
|
Aushon Biosystems Inc
03FC01FA |
6 Plex |
Each for $1,850.71
|
|
|||||
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Human | |
For simultaneous measurement of combinations of chemokines | |
Chemokine Array Kits |