Aushon Chemiluminescent Human Chemokine Array Kits
For simultaneous measurement of combinations of chemokines
Supplier: Aushon Biosystems Inc 03FC01FA
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Human | |
For simultaneous measurement of combinations of chemokines | |
Chemokine Array Kits | |
6 Plex |