Aushon Chemiluminescent Human Growth Factor Array Kits DFS Item
For simultaneous measurement of combinations of growth factors
Supplier:  Aushon Biosystems Inc 110081FA
 View more versions of this product
Catalog No. 110081FA
Explore available promotions
Description
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Specifications
Human
For simultaneous measurement of combinations of growth factors
1 Plex
SCF
Human Growth Factor Array Kits