missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aushon Chemiluminescent Human Growth Factor Array Kits DFS Item

For simultaneous measurement of combinations of growth factors

$876.65 - $876.65

Specifications

Reactivity Human
For Use With (Application) For simultaneous measurement of combinations of growth factors
Product Type Human Growth Factor Array Kits
Quantity 1 Plex
View More Specs

Products 2
Catalog Number Mfr. No. Detects Price Quantity  
Catalog Number Mfr. No. Detects Price Quantity  
110081FA DFS Item
View Documents
Aushon Biosystems Inc
110081FA
SCF
Each for $876.65
 
110101FA DFS Item
View Documents
Aushon Biosystems Inc
110101FA
TGFα
Each for $876.65
 
Description

Description

Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles

Specifications

Specifications

Human
For simultaneous measurement of combinations of growth factors
Human Growth Factor Array Kits
1 Plex
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'