Aushon Chemiluminescent Rat Cytokine Array Kits
For simultaneous measurement of combinations of cytokines
Supplier: Aushon Biosystems Inc 232921FA
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
For simultaneous measurement of combinations of Cytokines | |
Cytokine Array Kits | |
4 Plex |