Aushon Chemiluminescent Rat Cytokine Array Kits DFS Item
For simultaneous measurement of combinations of cytokines
$1461.10 - $2629.96
Specifications
For Use With (Application) For simultaneous measurement of combinations of Cytokines
Product Type Cytokine Array Kits
View More Specs
Products 6
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
232921FA DFS Item
View Documents
Aushon Biosystems Inc
232921FA
4 Plex
Each for $1,461.10
233F61FA DFS Item
View Documents
Aushon Biosystems Inc
233F61FA
8 Plex
Each for $2,240.35
232941FA DFS Item
View Documents
Aushon Biosystems Inc
232941FA
4 Plex
Each for $1,461.10
232B01FA DFS Item
View Documents
Aushon Biosystems Inc
232B01FA
4 Plex
Each for $1,461.10
233961FA DFS Item
View Documents
Aushon Biosystems Inc
233961FA
6 Plex
Each for $1,850.71
233FF1FA DFS Item
View Documents
Aushon Biosystems Inc
233FF1FA
10 Plex
Each for $2,629.96
Description
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Specifications
For simultaneous measurement of combinations of Cytokines
Cytokine Array Kits
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'