missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mopec Autopsy Saw Blades, SuperCut, Compatible with Stryker Saw
Mopec's exclusive titanium coating assures longer and sharper blade life. Fits all saws with radial mount arbors.
Supplier: Mopec BD116
Description
- Dimensions: 2.5 in. Diameter (64mm)
- Material: 420 Stainless steel with Titanium Nitride coating for added strength
- Fits Mopec Autopsy Saw (excludes Mopec Autopsy Saw 5000) and Stryker™ 810 Saw
Specifications
No | |
Autopsy Saw Blades, SuperCut, Compatible with Stryker Saw | |
Fits Mopec Autopsy Saw (excludes Mopec Autopsy Saw 5000) and Stryker™ Saw | |
2.5 in. |
Titanium Nitride Coating for added strength | |
Stainless Steel (420) | |
Round Blade |