Axygen™ 1.5 mL Conical Screw Cap Tubes
With chemical-resistant O-ring
Supplier:  Axygen™ SCT150OS
 View more versions of this product
Catalog No. 14-222-576
Explore available promotions
Description
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
  • Available as presterilized and nonsterile
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order
  • Caps available in range of colors for easy sample identification
Specifications
Specifications
RNase, DNase, pyrogen-free
Polypropylene
Polypropylene
O-ring
Screw Cap
20,000 x g
Orange
1.5 mL
Non-looped
Sterile
Conical
Product Suggestions
Product Suggestions