Axygen™ 1000 μL Universal Pipetter Tips: Bevelled
Compatible with most popular brands of pipettes
Supplier:  Axygen™ T1000BR
 View more versions of this product
Catalog No. 14-222-691
Description
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Beveled
Specifications
Specifications
100 to 1000 μL
Non-sterile
Autoclavable
Rack
Hinged rack
Polypropylene
Non-Filtered
Blue
DNase, RNase, and Pyrogen free
Beveled, Universal
Universal Fit