Axygen™ 1000 μL Universal Pipetter Tips: Bevelled
Compatible with most popular brands of pipettes
$102.09 - $223.78
Specifications
Volume (Metric) | 100 to 1000 μL |
---|---|
Autoclavable | Autoclavable |
Clean Claims | DNase, RNase, and Pyrogen free |
Tip Style | Beveled, Universal |
For Use With (Equipment) | Universal Fit |
Catalog Number | Mfr. No. | Color | Sterility | Format | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Sterility | Format | Price | Quantity | |||
14-222-690
![]() |
Axygen™
T1000B |
Blue | Non-sterile | Bulk |
Pack of 1000 Each for $102.09
Case of 5 Pack for $510.45
|
![]() |
|||
14-222-691
|
Axygen™
T1000BR |
Blue | Non-sterile | Rack |
Pack of 1000 Each for $195.17
Case of 5 Pack for $975.85
|
|
|||
14-222-692
|
Axygen™
T1000BRS |
Blue | Sterile | Rack |
Pack of 1000 Each for $214.78
Case of 5 Pack for $1,073.90
|
|
|||
14-222-693
|
Axygen™
T1000C |
Clear | Non-sterile | Bulk |
Pack of 1000 Each for $104.70
Case of 5 Pack for $523.50
|
|
|||
14-222-697
|
Axygen™
T1000CR |
Clear | Non-sterile | Rack |
Pack of 1000 Each for $189.91
Case of 5 Pack for $949.55
|
|
|||
14-222-698
|
Axygen™
T1000CRS |
Clear | Sterile | Rack |
Pack of 1000 Each for $223.78
Case of 5 Pack for $1,118.90
|
|
|||
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
- These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
- Nonpyrogenic
- Beveled
Specifications
100 to 1000 μL | |
DNase, RNase, and Pyrogen free | |
Universal Fit |
Autoclavable | |
Beveled, Universal | |
Polypropylene |