Axygen™ 200 μL Universal Pipetter Tips: 200 μL, Wide Bore
Designed to fit a wide variety of single and multi-channel pipettes
Supplier:  Axygen™ T205WBC
 View more versions of this product
Catalog No. 14-222-726
Explore available promotions
Description
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
(This is the suggested replacement for T-205-WB-C-L)
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore
Order Info
Available in bulk, racked or stack rack formats.
Specifications
Specifications
1 to 200 μL
Clear
DNase, RNase, and Pyrogen free
1,000 Tips/Pk., 20 Packs/Cs.
Polypropylene
Non-sterile
Autoclavable
Beveled, Wide Bore
Universal Fit Pipettor