Axygen™ Microplate Sealing Film and Tapes
Offer great economy for disposable ease.
Supplier:  Axygen™ PCRAS200
 View more versions of this product
Catalog No. 14-222-342
Explore available promotions
Description
Description
For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. 
Aluminum
  • Provides exceptional seal on multi-well plates with varied temperatures and reagents
  • With compression mats, films and tapes seal during thermocycling
  • Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
  • Pierceable, PCR-AS-200
Specifications
Specifications
Aluminum
5.74 in.
500/Cs.
5.74 x 3.11 in. (146 x 79.2 mm)
79.2 mm
Sealing Film
-80°C to +104°C
Non-sterile
146 mm
100/Pk.
3.11 in.
Designed for applications ranging from PCR and real-time PCR to ELISA and cell culture, ideal for light-sensitive samples
35 μm