Axygen™ Microplate Sealing Film and Tapes
Offer great economy for disposable ease.
$192.82 - $10683.00
Catalog Number | Mfr. No. | Material | Product Type | Dimensions (L x W) | Sterility | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Product Type | Dimensions (L x W) | Sterility | Price | Quantity | |||
50-400-316
|
Axygen™
BF400 |
Rayon | Breathable Sealing Film | 153 x 82.3 mm | Non-sterile |
Case for $3,912.79
|
|
|||
14-222-044
|
Axygen™
BF400S |
Rayon | Breathable Sealing Film | 153 x 82.3 mm | Sterile |
Case of 500 Each for $3,072.43
|
|
|||
14-222-131
|
Axygen™
HSFAS300L |
Aluminum | Sealing Film Roll | 115 x 500 m | Non-sterile |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
14-222-133
|
Axygen™
HSFTSL |
Polyester | CyclerSeal Sealing Film Roll | 115 x 500 m | Non-sterile |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
14-222-342
|
Axygen™
PCRAS200 |
Aluminum | Sealing Film | 146 x 79.2 mm | Non-sterile |
Pack of 100 Each for $367.00
Case of 5 Pack for $1,835.00
|
|
|||
14-222-343
|
Axygen™
PCRAS600 |
Aluminum | Sealing Film | 133.3 x 79.2 mm | Non-sterile |
Pack of 100 Each for $367.00
Case of 5 Pack for $1,835.00
|
|
|||
14-222-346
|
Axygen™
PCRSPS |
Polyester | AxySeal Sealing Film | 146 x 79.6 mm | Sterile |
Pack of 100 Each for $290.68
Case of 5 Pack for $1,453.40
|
|
|||
14-222-344
|
Axygen™
PCRSP |
Polyester | AxySeal Sealing Film | 146 x 79.6 mm | Non-sterile |
Pack of 100 Each for $192.82
Case of 5 Pack for $964.10
|
|
|||
14-222-347
|
Axygen™
PCRTS |
Polyester | CyclerSeal Sealing Film | 135.5 x 80 mm | Non-sterile |
Pack of 100 Each for $264.11
Case of 5 Pack for $1,320.55
|
|
|||
14-222-873
|
Axygen™
UC500 |
Polyolefin | UltraClear Sealing Film | Not Available | Non-sterile |
Pack of 100 Each for $882.61
Case of 5 Pack for $4,413.05
|
|
|||
Description
For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. Sealing film temperature ranges from -40°C to +104°C.
Breathable
- Allow effective gas exchange for cellular and bacterial cultivation — while preventing contamination
- Seal polypropylene and polystyrene culture plates, 96- and 384-well plates including other assay plates
- Provides exceptional seal on multi-well plates with varied temperatures and reagents
- With compression mats, films and tapes seal during thermocycling
- Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
- Pierceable, PCR-AS-200
- Exceptional seal across entire plate safeguards samples during transport or storage
- When used with Axygen's compression mat, this polyester film eliminates well-to-well contamination and cross-over in PCR applications
- Polyester-based with uniformly and consistently applied acrylic adhesive to eliminate the edge effect in sensitive ELISA assays
- Suitable for sealing tissue culture plates, short-term storage and incubation, transport and the containment of biohazardous solutions
- Available in both nonsterile and sterile packaging