Axygen™ PCR Tubes with 0.2 mL Dome Cap
Thin and consistent wall thickness for precise thermal transfer
Supplier:  Axygen™ PCR02DLC
 View more versions of this product
Catalog No. 50-400-526
Explore available promotions
Description
Description
Quantitative PCR compatible tubes for a wide range of Real-Time PCR instruments.
  • PCR tubes with 0.2mL volume
  • Tubes with caps sealed full-length; insertion cap ensures tight fit, eliminates sample evaporation
  • Choose from a wide variety of colors
Specifications
Specifications
RNase, DNase, pyrogen-free
0.2 mL
Polypropylene
Conical
Polypropylene
Tube
Clear
1000/Pk., 10Pk./Cs.
Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
Non-sterile
Domed
Maxymum Recovery, Domed Cap PCR Tubes