Axygen™ Presterilized Conical Centrifuge Tubes
Graduated tubes with conical bottom
Supplier: Axygen™ SCT15MLRS
Description
- Rated to 12,000 RCF
- Printed graduations and writing area for sample identification
- Screw caps can be closed with one hand.
- Sterile options are also available.
- RNase-/DNase-free
- Nonpyrogenic
Specifications
Screw Cap | |
15 mL | |
Polypropylene | |
Screw cap | |
Sterile | |
12,000 x g |
Not Autoclavable | |
Clear | |
For sample identification | |
50 Tubes and Caps/Unit, 20 Units/Cs. | |
Conical |