missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Axygen™ Screw Cap Tubes without Caps: Conical

Conical tubes

Supplier:  Axygen™ ST45

 View more versions of this product

Catalog No. 14-222-686


Explore available promotions

Description

Description

Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping

  • Tube volumes from 0.5 to 2.0mL
  • Choose clear or amber

Specifications

Specifications

RNase, DNase, pyrogen-free
Clear
500 Tubes/Pk., 8 Packs/Cs.
No
Conical
0.5 mL
Polypropylene
Non-sterile
Not Included
20,000 x g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions