missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Azetidine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5635103

 View more versions of this product

Catalog No. AAH5635103


Explore available promotions

Description

Description

Azetidine is involved in a high yielding palladium-catalyzed cross-coupling reaction with aryl bromides. Further, it is used in Ullmann type coupling reaction with iodonitroflourenes. In addition to this, it reacts with bis-(3-amino-propyl)-amine to prepare N,N'-bis-(3-amino-propyl)-propanediyldiamine using palladium as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azetidine is involved in a high yielding palladium-catalyzed cross-coupling reaction with aryl bromides. Further, it is used in Ullmann type coupling reaction with iodonitroflourenes. In addition to this, it reacts with bis-(3-amino-propyl)-amine to prepare N,N′-bis-(3-amino-propyl)-propanediyldiamine using palladium as a catalyst.

Solubility
Miscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Air sensitive and heat sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications

Specifications

Azetidine
0.847
−21°C (−6°F)
1.432
1 g
102384
azacyclobutane, trimethylene imine, 1,3-propylenimine, trimethylenimine, azetidin, unii-37s883xdwr, azetidine, l, polypropylene amine, azete, tetrahydro, azetidine
HONIICLYMWZJFZ-UHFFFAOYSA-N
azetidine
10422
57.1
503-29-7
61°C to 62°C
C3H7N
MFCD00005165
UN2924
Air and moisture sensitive
Miscible with water.
C1CNC1
57.096
CHEBI:30968
98%

Chemical Identifiers

503-29-7
57.096
HONIICLYMWZJFZ-UHFFFAOYSA-N
10422
azetidine
C3H7N
MFCD00005165
azacyclobutane, trimethylene imine, 1,3-propylenimine, trimethylenimine, azetidin, unii-37s883xdwr, azetidine, l, polypropylene amine, azete, tetrahydro, azetidine
CHEBI:30968
C1CNC1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

EINECSNumber : 207-963-8

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only