missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Azetidine, 98%, Thermo Scientific Chemicals

$109.05 - $1245.92

Chemical Identifiers

CAS 503-29-7
Molecular Formula C3H7N
Molecular Weight (g/mol) 57.096
MDL Number MFCD00005165
InChI Key HONIICLYMWZJFZ-UHFFFAOYSA-N
Synonym azacyclobutane, trimethylene imine, 1,3-propylenimine, trimethylenimine, azetidin, unii-37s883xdwr, azetidine, l, polypropylene amine, azete, tetrahydro, azetidine
PubChem CID 10422
ChEBI CHEBI:30968
IUPAC Name azetidine
SMILES C1CNC1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH56351MD
View Documents
Thermo Scientific Chemicals
H56351MD
250 mg
Each for $109.05
 
AAH5635103
View Documents
Thermo Scientific Chemicals
H5635103
1 g
Each for $280.79
 
AAH5635106
View Documents
Thermo Scientific Chemicals
H5635106
5 g
Each for $1,245.92
 
Description

Description

Azetidine is involved in a high yielding palladium-catalyzed cross-coupling reaction with aryl bromides. Further, it is used in Ullmann type coupling reaction with iodonitroflourenes. In addition to this, it reacts with bis-(3-amino-propyl)-amine to prepare N,N'-bis-(3-amino-propyl)-propanediyldiamine using palladium as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azetidine is involved in a high yielding palladium-catalyzed cross-coupling reaction with aryl bromides. Further, it is used in Ullmann type coupling reaction with iodonitroflourenes. In addition to this, it reacts with bis-(3-amino-propyl)-amine to prepare N,N′-bis-(3-amino-propyl)-propanediyldiamine using palladium as a catalyst.

Solubility
Miscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Air sensitive and heat sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications

Chemical Identifiers

503-29-7
57.096
HONIICLYMWZJFZ-UHFFFAOYSA-N
10422
azetidine
C3H7N
MFCD00005165
azacyclobutane, trimethylene imine, 1,3-propylenimine, trimethylenimine, azetidin, unii-37s883xdwr, azetidine, l, polypropylene amine, azete, tetrahydro, azetidine
CHEBI:30968
C1CNC1

Specifications

503-29-7
61°C to 62°C
C3H7N
MFCD00005165
UN2924
Air and moisture sensitive
Miscible with water.
C1CNC1
57.096
CHEBI:30968
98%
0.847
−21°C (−6°F)
1.432
250 mg
102384
azacyclobutane, trimethylene imine, 1,3-propylenimine, trimethylenimine, azetidin, unii-37s883xdwr, azetidine, l, polypropylene amine, azete, tetrahydro, azetidine
HONIICLYMWZJFZ-UHFFFAOYSA-N
azetidine
10422
57.1
Azetidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

EINECSNumber : 207-963-8

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only