Learn More
Bal Supply Stainless-Steel Pinch Clamps for Ball and Socket Joints
Easy, secure way to marry ball-and-socket joints
Supplier:  Bal Supply 20112WOS
 Afficher plus de versions de ce produit
Numéro de catalogue. 05-885B
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Spring-loaded pinch clamp with forked jaws on both sides of the joint ensures tight fit.
  • 05-885C and 05-885E through K have screw-locking device
  • Work well on both O-ring and ball-and-socket joints
  • Others are intended for ball-and-socket joints only
Spécifications
Spécifications
12/Pk.
Stainless Steel
18/7 to 18/9 Joints
Pinch Clamp
No Screw Locking Device
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS