missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Barium Chloride Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific FLB3410
Specifications
1560°C | |
962°C | |
White | |
Solid | |
≥99.0 % | |
MFCD00149154 | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
barium(2+) dihydrate dichloride | |
5284346 | |
244.26 | |
Certified ACS | |
14.0 to 16.0% | |
Decomposition Information: >100°C |
Barium Chloride Dihydrate | |
10326-27-9 | |
5 to 8 | |
10 kg | |
BaCl2H4O2 | |
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
O.O.[Cl-].[Cl-].[Ba++] | |
244.26 | |
CHEBI:86153 | |
≥99.0% | |
Pass Test | |
Poly Pail |
Chemical Identifiers
10326-27-9 | |
244.26 | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
5284346 | |
barium(2+) dihydrate dichloride |
BaCl2H4O2 | |
MFCD00149154 | |
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
CHEBI:86153 | |
O.O.[Cl-].[Cl-].[Ba++] |
Safety and Handling
DANGER!
Emergency Overview
Toxic if swallowed. Harmful by inhalation. May cause eye, skin, and respiratory tract irritation . May cause central nervous system effects.
NFPA
Health: 3
Flammability: 1
Instability: 0
SIGNAL WORD: DANGER!
DOTInformation : DOT Class 6.1, : Poison