missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
$303.42 - $3956.67
Chemical Identifiers
| CAS | 10326-27-9 |
|---|---|
| Molecular Formula | BaCl2H4O2 |
| Molecular Weight (g/mol) | 244.26 |
| MDL Number | MFCD00149154 |
| InChI Key | PWHCIQQGOQTFAE-UHFFFAOYSA-L |
| Synonym | barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d |
| PubChem CID | 5284346 |
| ChEBI | CHEBI:86153 |
| IUPAC Name | barium(2+) dihydrate dichloride |
| SMILES | O.O.[Cl-].[Cl-].[Ba++] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
B34-100
|
Thermo Fisher Scientific
FLB34100 |
100 g | Poly Bottle |
Each for $303.42
Case of 6 Each for $1,820.52
|
|
||||
B34-500
|
Thermo Fisher Scientific
FLB34500 |
500 g | Poly Bottle |
Each for $392.92
Case of 6 Each for $2,357.52
|
|
||||
|
B34-3
|
Thermo Fisher Scientific
FLB343 |
3 kg | Poly Bottle |
Each for $1,440.25
Case of 4 Each for $5,761.00
|
|
||||
|
B34-10
|
Fisher Chemical
FLB3410 |
10 kg | Poly Pail |
Each for $3,956.67
|
|
||||
Chemical Identifiers
| 10326-27-9 | |
| 244.26 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| 5284346 | |
| barium(2+) dihydrate dichloride |
| BaCl2H4O2 | |
| MFCD00149154 | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| CHEBI:86153 | |
| O.O.[Cl-].[Cl-].[Ba++] |
Specifications
| 1560°C | |
| 10326-27-9 | |
| 5 to 8 | |
| 100 g | |
| BaCl2H4O2 | |
| 1564 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| barium(2+) dihydrate dichloride | |
| 5284346 | |
| 244.26 | |
| Certified ACS | |
| 14.0 to 16.0% | |
| Decomposition Information: >100°C | |
| Barium Chloride Dihydrate |
| 962°C | |
| White | |
| Solid | |
| ≥99.0% | |
| MFCD00149154 | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| O.O.[Cl-].[Cl-].[Ba++] | |
| 244.26 | |
| CHEBI:86153 | |
| ≥99.0% | |
| Pass Test | |
| Poly Bottle | |
| 3.86g/mL |
Safety and Handling
DANGER!
Emergency Overview
Toxic if swallowed. Harmful by inhalation. May cause eye, skin, and respiratory tract irritation . May cause central nervous system effects.
NFPA
Health: 3
Flammability: 1
Instability: 0
SIGNAL WORD: DANGER!
DOTInformation : DOT Class 6.1, : Poison