missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Barium Chloride Dihydrate (Crystalline/Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific FLB34100

 View more versions of this product

Catalog No. B34-100



Explore available promotions

Description

Description

Specifications

Specifications

1560°C
962°C
White
Solid
≥99.0%
MFCD00149154
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
O.O.[Cl-].[Cl-].[Ba++]
244.26
CHEBI:86153
≥99.0%
Pass Test
Poly Bottle
3.86g/mL
Barium Chloride Dihydrate
10326-27-9
5 to 8
100 g
BaCl2H4O2
1564
PWHCIQQGOQTFAE-UHFFFAOYSA-L
barium(2+) dihydrate dichloride
5284346
244.26
Certified ACS
14.0 to 16.0%
Decomposition Information: >100°C

Chemical Identifiers

10326-27-9
244.26
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
barium(2+) dihydrate dichloride
BaCl2H4O2
MFCD00149154
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
CHEBI:86153
O.O.[Cl-].[Cl-].[Ba++]
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Toxic if swallowed. Harmful by inhalation. May cause eye, skin, and respiratory tract irritation . May cause central nervous system effects.
NFPA
Health: 3
Flammability: 1
Instability: 0

SIGNAL WORD: DANGER!

DOTInformation : DOT Class 6.1, : Poison

SDS Documents

Documents

Promotions

Promotions