Basix™ Microcentrifuge Tubes with Standard Snap Caps
Microtubes feature standard snap caps in choice of clear or multicolored vials
$40.14 - $52.79
Specifications
Max. RCF | 30,000 x g |
---|---|
Material | Polypropylene |
Graduated | Yes |
Closure Type | Snap Cap |
Catalog Number | Mfr. No. | Capacity (Metric) | Color | Graduations | Increments | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Color | Graduations | Increments | Price | Quantity | ||
02682000
|
Fisherbrand
502GRDSERV |
0.5 mL | Natural | 0.2 mL | 0.2, 0.4, 0.6mL |
Pack of 500 Each for $41.90
Case of 10 Pack for $419.00
|
|
||
02682001
|
Fisherbrand
502GRDMSERV |
0.5 mL | Assorted | 0.2 mL | 0.2, 0.4, 0.6mL |
Pack of 500 Each for $40.14
Case of 10 Pack for $401.40
|
|
||
02682002
|
Fisherbrand
509GRDSERV |
1.5 mL | Natural | 0.5 mL | 0.1, 0.5, 1.0, 1.5mL |
Pack of 500 Each for $50.69
Case of 10 Pack for $506.90
|
|
||
02682003
|
Fisherbrand
509GRDMSERV |
1.5 mL | Assorted | 0.5 mL | 0.1, 0.5, 1.0, 1.5mL |
Pack of 500 Each for $51.24
Case of 10 Pack for $512.40
|
|
||
02682004
|
Fisherbrand
508GRDSERV |
2 mL | Natural | 0.5 mL | 0.5, 1.0, 1.5, 2.0mL |
Pack of 500 Each for $51.93
Case of 10 Pack for $519.30
|
|
||
02682005
|
Fisherbrand
508GRDMSERV |
2 mL | Assorted | 0.5 mL | 0.5, 1.0, 1.5, 2.0mL |
Pack of 500 Each for $52.79
Case of 10 Pack for $527.90
|
|
||
Description
- Withstand forces up to 30,000 x g
- Temperature range of -80° to +121°C
- Available in either natural or mixed colors
- RNase, DNase-free
Specifications
30,000 x g | |
Polypropylene | |
Yes | |
Snap Cap |