BD BBL™ CultureSwab™ Sterile, Media-free Swabs
Sterile, media- and charcoal-free swabs for general specimen collection and transport
Supplier: BD 220115
Description
- Round-bottomed tube
- 5 1/4 in. Swab shaft with rayon-tipped swab
- Extra-large, preattached label
- Tamper-evident seal
- Lot number, expiration date and product description printed on the tube and on the peel pack
Specifications
Rayon | |
Round-bottom tube | |
5.25 in. |
Sterile | |
133.35mm | |
Single swab |