missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD BBL™ CultureSwab™ Sterile, Media-free Swabs

Sterile, media- and charcoal-free swabs for general specimen collection and transport

$124.16 - $124.16

Specifications

Tip Material Rayon
Sterility Sterile
Packaging Round-bottom tube
Length (Metric) Shaft 133.35mm
Length (English) Shaft 5.25 in.
View More Specs

Products 1
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
B4320115 Edge
View Documents
BD
220115
Single swab
Pack of 100 Each for $124.16
Edge
 
Description

Description

  • Round-bottomed tube
  • 5 1/4 in. Swab shaft with rayon-tipped swab
  • Extra-large, preattached label
  • Tamper-evident seal
  • Lot number, expiration date and product description printed on the tube and on the peel pack

Specifications

Specifications

Rayon
Round-bottom tube
5.25 in.
Sterile
133.35mm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'