Learn More
BD Acridine Orange Stain
For detecting microorganisms in direct smears by fluorescence
Supplier: BD 212537
Description
- Especially beneficial in screening normally sterile specimens where few organisms may be present
- Bacteria stain bright orange; human cells, tissue debris and proteinaceous material stain pale green to yellow
Specifications
| Acridine Orange Stain | |
| CE | |
| Acridine Orange |
| 4 x 250 mL | |
| Recommended for use in the fluorescent microscopic detection of microorganisms in direct smears prepared from clinical and non clinical materials |