BD Acridine Orange Stain
For detecting microorganisms in direct smears by fluorescence
$227.37 - $466.68
Specifications
Type | Acridine Orange Stain |
---|---|
Certifications/Compliance | CE |
For Use With (Application) | Recommended for use in the fluorescent microscopic detection of microorganisms in direct smears prepared from clinical and non clinical materials |
Stain Type | Acridine Orange |
Description
- Especially beneficial in screening normally sterile specimens where few organisms may be present
- Bacteria stain bright orange; human cells, tissue debris and proteinaceous material stain pale green to yellow
Specifications
Acridine Orange Stain | |
CE | |
Recommended for use in the fluorescent microscopic detection of microorganisms in direct smears prepared from clinical and non clinical materials | |
Acridine Orange |