missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Autoclip™ Wound Closing System
Ideal for use on lab animals or in veterinary surgery
Supplier: BD 427631
Description
Wound Clips
- 9mm nonsterile Autoclip wound clips 20 stainless-steel double clips per rack 100 clips (5 racks) per box, 10 boxes per case
Note: Items can be purchased as a kit or individually.
Specifications
Stainless Steel | |
Non-sterile | |
0.9 cm | |
For use on lab animals or in veterinary surgery |
Wound Clip | |
0.354 in. | |
100 Pk. | |
Yes |