Learn More
BD Bacto™ Dehydrated Agar
Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.
Supplier:  BD 214010
 View more versions of this product
Catalog No. DF0140-01-0
Only null left
Explore available promotions
Description
Description
Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.
  • Extraneous matter, pigmented portions and salts are reduced to minimum
  • Fine, light colored granules
  • Convenient for weighing and handling
Determines motility and growth of anaerobes and microaerophiles
Specifications
Specifications
ISO/DIS, 11014
Agar
Determination of motility and the growth of anaerobes and microaerophiles
454 g
1.5%
Dehydrated
Bottle
Product Suggestions
Product Suggestions