missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Bacto™ Dehydrated Agar
Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.
$319.87 - $16887.29
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Concentration | 1.5% |
Additive | Agar |
Form | Dehydrated |
For Use With (Application) | Determination of motility and the growth of anaerobes and microaerophiles |
Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity | |||||
DF0140-15-4
|
BD
214050 |
100 g | Bottle |
Each for $319.87
|
|
|||||
DF0140-01-0
|
BD
214010 |
454 g | Bottle |
Each for $819.37
|
|
|||||
DF0140-07-4
|
BD
214030 |
2000 g | Container |
Each for $2,938.24
|
|
|||||
DF0140-08-3
|
BD
214040 |
10,000 g | Pail |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
Description
Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.
- Extraneous matter, pigmented portions and salts are reduced to minimum
- Fine, light colored granules
- Convenient for weighing and handling
Determines motility and growth of anaerobes and microaerophiles
Specifications
ISO/DIS, 11014 | |
Agar | |
Determination of motility and the growth of anaerobes and microaerophiles |
1.5% | |
Dehydrated |