missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Bacto™ Yeast Extract

Gibco™ Bacto™ Dehydrated Culture Media Additive: Yeast Extract is supplied in its dehydrated form for the cultivation of heterotrophic microorganisms.

Supplier:  Gibco™ 212730

 View more versions of this product

Catalog No. DF0127-08-0


Explore available promotions

Description

Description

  • Bacto Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells, derived from primary grown baker's yeast
  • These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
  • Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation
  • Bacto Yeast Extract is one of the most complete and versatile fermentation supplements available
  • It is an important ingredient for the microbiological assay of vitamins
  • Yeast extract is also of value in the assay of antibiotics
  • B factor, a growth substance necessary for the production of rifampin in a Nocardia species, can be isolated from yeast extract
  • Bacto Yeast Extract, Technical, was developed to provide products priced for the biotechnology market with acceptable clarity and growth-promoting characteristics
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C

Related yeast extract products include:

  • Gibco Yeast Extract – Developed to provide a product for the biotechnology/pharmaceutical market with acceptable clarity and growth-promoting characteristics. Media formulations containing yeast extract are specified in standard methods for various applications.
  • Difco Yeast Extract UF – Ultra-filtered and specifically designed for mammalian cell culture applications. With its low endotoxin level and high content of naturally occurring B vitamins, it can be used effectively to reduce the need for fetal bovine serum. It has an endotoxin level of less than or equal to 500 EU/g
  • Difco Yeast Extract Low-Dusting (LD) – Designed to limit the amount of dust generated compared to other spray-dried yeast extracts
Specifications

Specifications

Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Bacterial Media
Formulation of culture media for the cultivation of heterotrophic microorganisms
10 kg
Mammalian, Insect, Microbial
Pail
Bacto™
Yeast, Animal Origin-Free
Videos
SDS
Documents

Documents

Promotions

Promotions