missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Bacto™ Yeast Extract
Gibco™ Bacto™ Dehydrated Culture Media Additive: Yeast Extract is supplied in its dehydrated form for the cultivation of heterotrophic microorganisms.
$422.00 - $22150.91
Specifications
Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
---|---|
Culture Type | Mammalian, Insect, Microbial |
For Use With (Application) | Formulation of culture media for the cultivation of heterotrophic microorganisms |
Product Line | Bacto™ |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
DF0127-17-9
|
Gibco™
212750 |
500 g |
Each for $422.00
|
|
|||||
DF0127-07-1
|
Gibco™
212720 |
2 kg |
Each for $1,381.00
|
|
|||||
DF0127-08-0
|
Gibco™
212730 |
10 kg |
Each for $6,811.34
|
|
|||||
212710
|
Gibco™
212710 |
50 kg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Bacto Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells, derived from primary grown baker's yeast
- These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
- Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation
- Bacto Yeast Extract is one of the most complete and versatile fermentation supplements available
- It is an important ingredient for the microbiological assay of vitamins
- Yeast extract is also of value in the assay of antibiotics
- B factor, a growth substance necessary for the production of rifampin in a Nocardia species, can be isolated from yeast extract
- Bacto Yeast Extract, Technical, was developed to provide products priced for the biotechnology market with acceptable clarity and growth-promoting characteristics
- Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Related yeast extract products include:
- Gibco Yeast Extract – Developed to provide a product for the biotechnology/pharmaceutical market with acceptable clarity and growth-promoting characteristics. Media formulations containing yeast extract are specified in standard methods for various applications.
- Difco Yeast Extract UF – Ultra-filtered and specifically designed for mammalian cell culture applications. With its low endotoxin level and high content of naturally occurring B vitamins, it can be used effectively to reduce the need for fetal bovine serum. It has an endotoxin level of less than or equal to 500 EU/g
- Difco Yeast Extract Low-Dusting (LD) – Designed to limit the amount of dust generated compared to other spray-dried yeast extracts
Specifications
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. | |
Mammalian, Insect, Microbial | |
Formulation of culture media for the cultivation of heterotrophic microorganisms | |
Bacto™ |