missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: XLD Agar
Selective and differential media for isolation and differentiation of enteric pathogens, especially Shigella and Salmonella species
Supplier: BD 221284
Description
- Ready-to-use
- Inhibitory to gram-positive organisms
- 100 x 10mm and 100 x 15mm-style plates
Specifications
| ISO/DIS, 11014 | |
| 2°C to 8°C | |
| United States | |
| Xylose 3.5g/L, L-Lysine 5g/L, Lactose 7.5g/L, Saccharose 7.5g/L, Sodium Chloride 5g/L, Yeast extract 3g/L, Phenol red 0.08g/L, Sodium Desoxycholate 2.5g/L, Sodium Thiosulfate 6.8g/L, Ferric Ammonium Citrate 0.8g/L, Agar 13.5g/L | |
| Plate |
| Light Orange | |
| Liquid | |
| XLD Agar | |
| Enteric pathogens, especially Shigella species | |
| 100/Pk. |