missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD BBL™ Prepared Plated Media: XLD Agar
Selective and differential media for isolation and differentiation of enteric pathogens, especially Shigella and Salmonella species
$217.70 - $217.70
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Color | Light Orange |
Content And Storage | 2°C to 8°C |
Form | Liquid |
Manufacturing Origin | United States |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
B4321284
|
BD
221284 |
100/Pk. |
Pack of 100 Each for $217.70
|
|
||||||
Description
- Ready-to-use
- Inhibitory to gram-positive organisms
- 100 x 10mm and 100 x 15mm-style plates
Specifications
ISO/DIS, 11014 | |
2°C to 8°C | |
United States | |
Xylose 3.5g/L, L-Lysine 5g/L, Lactose 7.5g/L, Saccharose 7.5g/L, Sodium Chloride 5g/L, Yeast extract 3g/L, Phenol red 0.08g/L, Sodium Desoxycholate 2.5g/L, Sodium Thiosulfate 6.8g/L, Ferric Ammonium Citrate 0.8g/L, Agar 13.5g/L | |
Plate |
Light Orange | |
Liquid | |
XLD Agar | |
Enteric pathogens, especially Shigella species |