BD BBL™ Replacement Reagents for Gram Stain Kits
For BBL Gram Stain Kits
Supplier: BD 212531
Description
- Complete stain kits reduce the mess associated with Gram staining procedures
- Break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills
- Color-coded labels allow quick identification of reagents
- Stabilized Iodine Solution requires no mixing and has a long shelf life
Order Info
Available in packs of four 250mL bottles or individual one-gallon bottles
Specifications
Safranin | |
CE | |
15° to 30°C |
4 x 250mL | |
Used to stain microorganisms from cultures or specimens by the differential gram method | |
Safranin |