BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Ciprofloxacin31
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
Supplier:  BD 231658
 View more versions of this product
Catalog No. B31658
Explore available promotions
Description
Description
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Specifications
Ciprofloxacin31
5μg
Disc
10 Cartridges (50 discs/cartridge)
Videos
SDS
Documents
Documents
Accessories
Promotions
Promotions
For in vitro diagnostic use